Learn More
Sigma Aldrich Fine Chemicals Biosciences LifeSep magnetic separator1EA
Catalog No. 502909038
Encompass
Durable easy-to-use and designed to accommodate industry standard microtitre plates. For use in cell sorting RNA and DNA isolation and purification of biomolecules. Fast and easy aspiration is achieved with our Neodymium-Iron-Boron (Nd-Fe-B) magnet configuration.Patented Nd-Fe-B magnet configuration for optimal resultsOptimized field gradient within wellsRapid separation of micro- and nanometer beadsEasy aspirationSuperior particle retentionRemovable base for easy cleaning
Catalog No. 50-290-9038 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7401581EA
Durable easy-to-use and designed to accommodate industry standard microtitre plates. For use in cell sorting RNA and DNA isolation and purification of biomolecules. Fast and easy aspiration is achieved with our Neodymium-Iron-Boron (Nd-Fe-B) magnet configuration.Patented Nd-Fe-B magnet configuration for optimal resultsOptimized field gradient within wellsRapid separation of micro- and nanometer beadsEasy aspirationSuperior particle retentionRemovable base for easy cleaning