Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LifeSign Easicult™ TTC
SDP

Catalog No. 23264778 Shop All Lifesign LLC Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Form:
Solid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
23-264778 Solid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-264778 Supplier LifeSign Supplier No. 4030
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dip-slide method designed for monitoring microbes in industrial fluids

  • Prolongs lifetime of industrial fluids by allowing adjustment of microbial level
  • Prevents microbial matter from clogging filters and pumps
  • Prevents machine, tank, and pipe corrosion by minimizing amount of microbial metabolites and by stabilizing amount of lubrication
  • Optimizes level of biocides by monitoring microbial counts
  • Prevents process stoppages caused by microbes
  • Improves hygienic conditions of working areas

Specifications

Form Solid
Manufacturing Origin United States
For Use With (Application) For monitoring microbes in industrial fluids
Packaging Type Slide
Quantity 10 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.