Learn More
LifeSign Lifesign MI™ Cardiac Markers Test Kit
SDP
Catalog No. 23264799 Shop All Lifesign LLC Products
Click to view available options
Detectable Analytes:
Myoglobin, Troponin I, CK-MB
Troponin I
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes
23-264-799 Troponin I
23-264-800 Myoglobin, Troponin I, CK-MB
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-264-799 Supplier LifeSign Supplier No. 60101
Add to Cart
Add to Cart
Lifesign MI™ tests are for rapid detection of cardiac markers
LifeSign MI cardiac tests allow for the rapid qualitative detection of cardiac markers in 15 minutes to aid in the diagnosis of acute myocardial infarction. An easy to read immunoassay that requires only three drops of whole blood, serum or plasma.
Performing these tests at the point of care:
  • Improves patient care and the efficiency of chest pain management, treatment and triage decisions
  • Enables prompt emergency room decisions, thereby reducing unnecessary testing and admissions
  • Simple one-step testing
  • Multiple panel assays
  • Easy-to-read results with distinct bands and a clear background
LifeSign MI cardiac tests allow for the rapid qualitative detection of cardiac markers in 15 minutes to aid in the diagnosis of acute myocardial infarction.  An easy to read immunoassay that requires only three drops of whole blood, serum or plasma.
Performing these tests at the point of care:
  • Improves patient care and the efficiency of chest pain management, treatment and triage decisions
  • Enables prompt emergency room decisions, thereby reducing unnecessary testing and admissions
  • Simple one-step testing
  • Multiple panel assays
  • Easy-to-read results with distinct bands and a clear background
Specifications
Detectable Analytes Troponin I
Sample Type Whole blood, Serum, Plasma
Type Cardiac Marker Test
Quantity 20 Tests