Learn More
LifeSign Uricult™ Media
SDP
Catalog No. 23264782 Shop All Lifesign LLC Products
Click to view available options
Incubation Time:
24 hr.
Catalog No. Incubation Time
23-264-782 24 hr.
1 options
Catalog No. 23-264-782 Supplier LifeSign Supplier No. 1003
Selective media for monitoring the growth of gram negative bacteria in industrial fluids
The addition of Polymixin in the CLED agar creates a selective medium for the growth of Enterococci, Staphlococci, Proteus, and Yeast while preventing the growth of most E. coli, Klebsiella, and Enterobacter. The neutral red indicator included in the media causes the colonies of lactose fermenting bacteria to appear pink or red.
Specifications
Product Type Microbiology Media
For Use With (Application) Monitoring microbes in industrial fluids
Incubation Time 24 hr.
Packaging Type Dip Slide