Learn More
Sigma Aldrich Lifesupport™ Support Slurry For Fresh Bioprinting
Catalog No. 9154671EA Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
91-546-71EA 1 Ea.
1 options
Catalog No. 91-546-71EA Supplier Sigma Aldrich Supplier No. 9154671EA
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
LifeSupport(TM) is the support slurry for FRESH bioprinting. FRESH stands for Freeform Reversible Embedding of Suspended Hydrogels. It is a 3D bioprinting technique using a thermoreversible support bath to enable deposition of hydrogels in complex 3D structure. Each LifeSupport(TM) printing kit comes with 4 units of 2 grams of sterile LifeSupport(TM) powder. One gram of powder hydrates to approximately 10 mL of FRESH support material.LifeSupport is a trademark of Advanced BioMatrix, Inc.
Specifications
Quantity 1 Ea.