Learn More
ACROBiosystems Recombinant Protein;Mouse LIF Protein;HEK293;50UG;LIF,LIF,CDF,DIA,HILDA,MLPLI
Catalog No. 501658548
Encompass_Preferred
Recombinant Protein;Mouse LIF Protein;50UG;Mouse LIF, His Tag (LIF-M5227) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Phe 203 (Accession # P09056).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 20.7 kDa. The protein migrates as 33-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8548 Supplier ACROBiosystems Supplier No. LIFM522750UG
Recombinant Protein;Mouse LIF Protein;50UG;Mouse LIF, His Tag (LIF-M5227) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Phe 203 (Accession # P09056).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 20.7 kDa. The protein migrates as 33-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.