Learn More
Medchemexpress LLC Lificiguat | 170632-47-0 | 99.7% | 304.34 | 200 MG
SDP
Catalog No. 5000359535
Encompass_Preferred
Lificiguat (YC-1) is a soluble guanylyl cyclase (sGC) activator that binds to the β subunit of sGC. It stimulates sGC activity, especially in the presence of CO or NO, leading to several hundred-fold activation. This compound has also been shown to inhibit tumor growth and improve rodent learning behavior.
  • Binds to the β subunit of soluble guanylyl cyclase (sGC)
  • Activates sGC, synergistically with carbon monoxide or nitric oxide
  • Inhibits tumor growth
  • Improves rodent learning behavior
  • Enhances long-term potentiation (LTP)
Catalog No. 50-003-59535 Supplier Medchemexpress LLC Supplier No. HY14927200MG
Lificiguat (YC-1) is a soluble guanylyl cyclase (sGC) activator that binds to the β subunit of sGC. It stimulates sGC activity, especially in the presence of CO or NO, leading to several hundred-fold activation. This compound has also been shown to inhibit tumor growth and improve rodent learning behavior.
  • Binds to the β subunit of soluble guanylyl cyclase (sGC)
  • Activates sGC, synergistically with carbon monoxide or nitric oxide
  • Inhibits tumor growth
  • Improves rodent learning behavior
  • Enhances long-term potentiation (LTP)