Learn More
Medchemexpress LLC HY-18957 5mg Medchemexpress, Lifirafenib CAS:1446090-79-4 Purity:>98%
SDP
Catalog No. 502082099
Encompass_Preferred
Medchemexpress, HY-18957 5mg Lifirafenib CAS:1446090-79-4 Lifirafenib (BGB-283) is a novel and potent Raf Kinase and EGFR inhibitor with IC50 values of 23 and 29 nM for recombinant BRafV600E and EGFR, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2099 Supplier Medchemexpress LLC Supplier No. HY189575MG
Medchemexpress, HY-18957 5mg Lifirafenib CAS:1446090-79-4 Lifirafenib (BGB-283) is a novel and potent Raf Kinase and EGFR inhibitor with IC50 values of 23 and 29 nM for recombinant BRafV600E and EGFR, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.