Learn More
Medchemexpress LLC HY-19344 10mg Medchemexpress, Lifitegrast CAS:1025967-78-5 Purity:>98%
SDP
Catalog No. 502082121
Encompass_Preferred
Medchemexpress, HY-19344 10mg Lifitegrast CAS:1025967-78-5 Lifitegrast (SAR 1118) is an integrin lymphocyte function-associated antigen-1 (LFA-1) antagonist; inhibits Jurkat T cell attachment to ICAM-1 with an IC50 of 2.98 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2121 Supplier Medchemexpress LLC Supplier No. HY1934410MG
Add to Cart
Add to Cart
Medchemexpress, HY-19344 10mg Lifitegrast CAS:1025967-78-5 Lifitegrast (SAR 1118) is an integrin lymphocyte function-associated antigen-1 (LFA-1) antagonist; inhibits Jurkat T cell attachment to ICAM-1 with an IC50 of 2.98 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.