Learn More
TARGETMOL CHEMICALS INC LIFITEGRAST 50MG
SDP
Catalog No. 502580033
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Lifitegrast (SAR 1118) is a lymphocyte function-associated antigen-1 antagonist. purity: 99%
Catalog No. 50-258-0033 Supplier TARGETMOL CHEMICALS INC Supplier No. T397750MG
Also available in 5 mg 10 mg 25 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Lifitegrast (SAR 1118) is a lymphocyte function-associated antigen-1 antagonist. purity: 99%