Learn More
Medchemexpress LLC LIFR Human HEK293 50ug | 50ug
SDP
Catalog No. 5000186339
Encompass_Preferred
LIFR Protein a signal-transducing molecule potentially shares a common pathway with IL6ST Its soluble form inhibits LIF binding to target cell receptors reducing biological activity LIFR forms a heterodimer with IL6ST interacting with the CNTF and CNTFR complex implying potential crosstalk and collaborative actions in cellular processes LIFR Protein Human (HEK293 Fc) is the recombinant human-derived LIFR protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-86339 Supplier Medchemexpress LLC Supplier No. HYP7705850UG
LIFR Protein a signal-transducing molecule potentially shares a common pathway with IL6ST Its soluble form inhibits LIF binding to target cell receptors reducing biological activity LIFR forms a heterodimer with IL6ST interacting with the CNTF and CNTFR complex implying potential crosstalk and collaborative actions in cellular processes LIFR Protein Human (HEK293 Fc) is the recombinant human-derived LIFR protein expressed by HEK293 with C-hFc labeled tag