Learn More
Medchemexpress LLC LIFR Mouse HEK293 5ug | 5ug
SDP
Catalog No. 5000186186
Encompass_Preferred
LIFR Protein a signal-transducing molecule may share a common pathway with IL6ST Its soluble form acts as an inhibitor blocking LIF binding to receptors and reducing biological activity LIFR forms a heterodimer with IL6ST interacting with the CNTF and CNTFR complex indicating potential cross-talk and cooperative actions in cellular processes LIFR Protein Mouse (HEK293 His) is the recombinant mouse-derived LIFR protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-86186 Supplier Medchemexpress LLC Supplier No. HYP770615UG
LIFR Protein a signal-transducing molecule may share a common pathway with IL6ST Its soluble form acts as an inhibitor blocking LIF binding to receptors and reducing biological activity LIFR forms a heterodimer with IL6ST interacting with the CNTF and CNTFR complex indicating potential cross-talk and cooperative actions in cellular processes LIFR Protein Mouse (HEK293 His) is the recombinant mouse-derived LIFR protein expressed by HEK293 with C-His labeled tag