Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC LIFR protein, rat (HEK293, His) | >95.0% | 50 UG
SDP

Catalog No. 5000292356
Encompass_Preferred

LIFR protein, rat is a recombinant, glycosylated form of the leukemia inhibitory factor receptor expressed in HEK293 mammalian cells and supplied as a lyophilized His-tagged protein for research use. It is designed for biochemical and cell-based assays, receptor binding and signaling studies, and antibody validation, with high purity and low endotoxin to support sensitive experiments.

  • Expressed in HEK293 mammalian cells, preserving native post-translational modifications
  • Contains a C-terminal His tag for detection and purification
  • Glycosylated protein with an approximate molecular weight of 116 kDa
  • Lyophilized formulation for convenient storage and transport
  • Purity greater than 95% by reducing SDS-PAGE
  • Endotoxin level <1 EU/μg suitable for cell-based work
  • Recommended storage at -20°C; aliquot to avoid freeze-thaw cycles

Catalog No. 50-002-92356 Supplier Medchemexpress LLC Supplier No. HYP7590750UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LIFR protein, rat is a recombinant, glycosylated form of the leukemia inhibitory factor receptor expressed in HEK293 mammalian cells and supplied as a lyophilized His-tagged protein for research use. It is designed for biochemical and cell-based assays, receptor binding and signaling studies, and antibody validation, with high purity and low endotoxin to support sensitive experiments.

  • Expressed in HEK293 mammalian cells, preserving native post-translational modifications
  • Contains a C-terminal His tag for detection and purification
  • Glycosylated protein with an approximate molecular weight of 116 kDa
  • Lyophilized formulation for convenient storage and transport
  • Purity greater than 95% by reducing SDS-PAGE
  • Endotoxin level <1 EU/μg suitable for cell-based work
  • Recommended storage at -20°C; aliquot to avoid freeze-thaw cycles

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.