Learn More
MSC Lift-All Chain Sling, Control Plate & Grab Hook, 6 ft Long, 2700 lb Capacity, Alloy Steel
Catalog No. 5000516034
Encompass
This chain sling features a control plate and grab hook, designed for high-speed applications. It offers a 6 ft length and a vertical capacity of 2700 lb. Made from durable alloy steel with a 7/32 inch chain, it is suitable for various lifting tasks.
  • Sling grade 100
  • Hook opening of 0.85 inches
  • Capacity at 60 degrees of 4700 lbs
  • Powder-coated finish in yellow and natural colors
  • Abrasion-resistant design
  • Adheres to ASME B30.9, OSHA, and NACM standards
Catalog No. 50-005-16034 Supplier MSC Supplier No. 89751705
This chain sling features a control plate and grab hook, designed for high-speed applications. It offers a 6 ft length and a vertical capacity of 2700 lb. Made from durable alloy steel with a 7/32 inch chain, it is suitable for various lifting tasks.
  • Sling grade 100
  • Hook opening of 0.85 inches
  • Capacity at 60 degrees of 4700 lbs
  • Powder-coated finish in yellow and natural colors
  • Abrasion-resistant design
  • Adheres to ASME B30.9, OSHA, and NACM standards