Learn More
MSC Lift-All Chain Sling, Double-Leg, 5 ft., 2700 lb Capacity
Catalog No. 5000530155
Encompass
This double-leg chain sling measures 5 feet long and is designed for heavy-duty lifting applications. It offers a vertical capacity of 2700 lb and is constructed from durable alloy steel for both the chain and hardware, ensuring reliability and longevity.
  • Double-leg configuration with an oblong master link and sling hooks
  • Measures 5 ft. long
  • Provides a vertical lifting capacity of 2700 lb
  • Features 7/32 in. alloy steel chain
  • Abrasion and corrosion resistant
  • Meets ASME B30.9, OSHA, and NACM standards
Catalog No. 50-005-30155 Supplier MSC Supplier No. 56443310
This double-leg chain sling measures 5 feet long and is designed for heavy-duty lifting applications. It offers a vertical capacity of 2700 lb and is constructed from durable alloy steel for both the chain and hardware, ensuring reliability and longevity.
  • Double-leg configuration with an oblong master link and sling hooks
  • Measures 5 ft. long
  • Provides a vertical lifting capacity of 2700 lb
  • Features 7/32 in. alloy steel chain
  • Abrasion and corrosion resistant
  • Meets ASME B30.9, OSHA, and NACM standards