Learn More
MSC Lift-All Chain Sling, Double-Leg, Alloy Steel, 5 ft Length, 7400 lb Working Load Limit
Catalog No. 5000534709
Encompass
This double-leg chain sling features an oblong master link and sling hooks, making it suitable for a variety of lifting applications. Constructed from alloy steel for both the chain and hardware, it offers excellent durability and resistance to abrasion and corrosion. Designed for demanding industrial environments, this sling meets ASME B30.9, OSHA, and NACM standards, ensuring safety and compliance.
  • Double-leg configuration
  • Oblong master link
  • Sling hooks included
  • 5 ft length
  • Alloy steel chain and hardware material
  • 7400 lb working load limit
  • 9/32 in chain size
  • Grade 100 chain
  • Powder-coated natural finish
  • Abrasion-resistant
  • Corrosion-resistant
  • Meets ASME B30.9, OSHA, and NACM standards
  • 1 3/16 in hook opening
  • Maximum operating temperature of 400°F
  • Fixed chain adjustability
Catalog No. 50-005-34709 Supplier MSC Supplier No. 69257079
This double-leg chain sling features an oblong master link and sling hooks, making it suitable for a variety of lifting applications. Constructed from alloy steel for both the chain and hardware, it offers excellent durability and resistance to abrasion and corrosion. Designed for demanding industrial environments, this sling meets ASME B30.9, OSHA, and NACM standards, ensuring safety and compliance.
  • Double-leg configuration
  • Oblong master link
  • Sling hooks included
  • 5 ft length
  • Alloy steel chain and hardware material
  • 7400 lb working load limit
  • 9/32 in chain size
  • Grade 100 chain
  • Powder-coated natural finish
  • Abrasion-resistant
  • Corrosion-resistant
  • Meets ASME B30.9, OSHA, and NACM standards
  • 1 3/16 in hook opening
  • Maximum operating temperature of 400°F
  • Fixed chain adjustability