Learn More
MSC Lift-All Endless Round Sling, 8 ft, 16800 lb Vertical Capacity, Polyester, White
Catalog No. 5000531910
Encompass
This endless round sling is constructed from polyester yarn within a double-wall tubular jacket. Its design allows core yarns to move independently, preventing strength loss from abrasion and enabling wear points to be shifted, which extends the sling's life. It resists UV sunlight, grease, and most acids.
  • Color coded capacities for quick identification.
  • Provides serial numbered identification for traceability.
  • Low stretch (about 3% at rated capacity).
  • Maintains 100% of original strength until jacket damage exposes core yarns.
  • When core yarns are visible, sling must be removed from service.
  • Chokes tighter and releases easier than other slings.
  • Good for low headroom lifts.
  • Light weight reduces fatigue and strain on riggers.
  • Complies with ASME B30.9, OSHA, and WSTDA standards.
  • Operating temperature range of -60 to 200 degrees F.
Catalog No. 50-005-31910 Supplier MSC Supplier No. 89673917
This endless round sling is constructed from polyester yarn within a double-wall tubular jacket. Its design allows core yarns to move independently, preventing strength loss from abrasion and enabling wear points to be shifted, which extends the sling's life. It resists UV sunlight, grease, and most acids.
  • Color coded capacities for quick identification.
  • Provides serial numbered identification for traceability.
  • Low stretch (about 3% at rated capacity).
  • Maintains 100% of original strength until jacket damage exposes core yarns.
  • When core yarns are visible, sling must be removed from service.
  • Chokes tighter and releases easier than other slings.
  • Good for low headroom lifts.
  • Light weight reduces fatigue and strain on riggers.
  • Complies with ASME B30.9, OSHA, and WSTDA standards.
  • Operating temperature range of -60 to 200 degrees F.