Learn More
MSC Lift-All Endless Round Sling, Polyester, 12 ft, 21,200 lb, Blue
Catalog No. 5000513042
Encompass
This polyester endless round sling is designed for versatile industrial lifting. Its continuous loop construction with a double-wall tubular jacket allows core yarns to move independently, enhancing load stability and extending product lifespan by enabling wear points to be shifted. It meets or exceeds OSHA and ASME B30.9 standards.
  • Achieves load stability and balance by spreading sling legs.
  • Extends sling life by allowing wear points to be shifted.
  • Lightweight and pliable for easy rigging and storage.
  • Synthetic materials are gentle on hands and loads.
  • Low stretch (approx. 3% at rated capacity) for stable lifts.
  • Conforms securely to load shapes.
  • Features red core yarns for visual warning of damage.
  • Identification tag for traceability and OSHA compliance.
Catalog No. 50-005-13042 Supplier MSC Supplier No. 74698804
This polyester endless round sling is designed for versatile industrial lifting. Its continuous loop construction with a double-wall tubular jacket allows core yarns to move independently, enhancing load stability and extending product lifespan by enabling wear points to be shifted. It meets or exceeds OSHA and ASME B30.9 standards.
  • Achieves load stability and balance by spreading sling legs.
  • Extends sling life by allowing wear points to be shifted.
  • Lightweight and pliable for easy rigging and storage.
  • Synthetic materials are gentle on hands and loads.
  • Low stretch (approx. 3% at rated capacity) for stable lifts.
  • Conforms securely to load shapes.
  • Features red core yarns for visual warning of damage.
  • Identification tag for traceability and OSHA compliance.