Learn More
MSC Lift-All Endless Round Sling, Polyester, 8 ft, 8400 lb Vertical Capacity, Yellow
Supplier: MSC 89673735
This endless round sling is constructed from polyester, offering durability and flexibility for various lifting tasks. With an 8 ft length and an 8400 lb vertical capacity, it is designed to promote safety and efficiency in operations. The sling also features abrasion resistance to extend its service life.
- Load stability and balance can be achieved by spreading sling legs.
- Wear points can be shifted to extend sling life.
- Most flexible style of sling.
- Individual slings can be attached together using appropriate hardware.
- Abrasion-resistant.