Learn More
MSC Lift-All Eye & Eye Sling, Nylon, 2 in x 3 ft, 12800 lb Basket Capacity
Catalog No. 5000527620
Encompass
This Eye & Eye Sling is a Flat Eye & Eye, Type 3 configuration, made from durable nylon. It measures 2 in wide by 3 ft long, offering a 6,400 lb vertical capacity, 5,000 lb choker capacity, and 12,800 lb basket capacity. With two plies for strength and abrasion resistance, it meets WSTDA, ASME B30.9, and OSHA 1910.184 standards, operating up to 200°F.
  • Flat Eye & Eye, Type 3 configuration.
  • Durable nylon material.
  • 2 in width, 3 ft length.
  • 6,400 lb vertical capacity.
  • 5,000 lb choker capacity.
  • 12,800 lb basket capacity.
  • Two plies for enhanced strength.
  • Abrasion-resistant.
  • 10 in eye length.
  • High-visibility yellow color.
  • Meets WSTDA, ASME B30.9, and OSHA 1910.184 standards.
  • Maximum operating temperature of 200°F.
  • Red core yarns signal damage for inspection.
Catalog No. 50-005-27620 Supplier MSC Supplier No. 81194003
This Eye & Eye Sling is a Flat Eye & Eye, Type 3 configuration, made from durable nylon. It measures 2 in wide by 3 ft long, offering a 6,400 lb vertical capacity, 5,000 lb choker capacity, and 12,800 lb basket capacity. With two plies for strength and abrasion resistance, it meets WSTDA, ASME B30.9, and OSHA 1910.184 standards, operating up to 200°F.
  • Flat Eye & Eye, Type 3 configuration.
  • Durable nylon material.
  • 2 in width, 3 ft length.
  • 6,400 lb vertical capacity.
  • 5,000 lb choker capacity.
  • 12,800 lb basket capacity.
  • Two plies for enhanced strength.
  • Abrasion-resistant.
  • 10 in eye length.
  • High-visibility yellow color.
  • Meets WSTDA, ASME B30.9, and OSHA 1910.184 standards.
  • Maximum operating temperature of 200°F.
  • Red core yarns signal damage for inspection.