Learn More
MSC Lift-All Eye & Eye Web Sling, 4 ft Long, 1 in Wide, 3200 lb Vertical Capacity, Polyester
Catalog No. 5000531302
Encompass
This Eye & Eye web sling is engineered for reliable lifting applications. It features a specific length and width, along with a significant vertical lifting capacity, making it suitable for various industrial and commercial tasks. The durable polyester construction ensures long-lasting performance.
  • Eye & Eye sling type
  • Polyester material
  • 4 ft long
  • 1 in wide
  • 3200 lb vertical capacity
  • 2500 lb choker capacity
  • 6400 lb basket capacity
  • 2 plies
  • 9 in eye length
  • Abrasion-resistant
  • Meets WSTDA, ASME B30.9, and OSHA 1910.184 standards
Catalog No. 50-005-31302 Supplier MSC Supplier No. 81193450
This Eye & Eye web sling is engineered for reliable lifting applications. It features a specific length and width, along with a significant vertical lifting capacity, making it suitable for various industrial and commercial tasks. The durable polyester construction ensures long-lasting performance.
  • Eye & Eye sling type
  • Polyester material
  • 4 ft long
  • 1 in wide
  • 3200 lb vertical capacity
  • 2500 lb choker capacity
  • 6400 lb basket capacity
  • 2 plies
  • 9 in eye length
  • Abrasion-resistant
  • Meets WSTDA, ASME B30.9, and OSHA 1910.184 standards