Learn More
MSC Lift-All Eye & Eye Wire Rope Sling, 5/8 in x 6 ft, 6x19 IWRC, Steel
Catalog No. 5000529699
Encompass
This eye and eye wire rope sling features a 5/8 in diameter and a 6 ft length, ideal for various lifting operations. It is constructed from 6x19 IWRC EIPS steel, providing excellent abrasion resistance and value. The sling is equipped with a mechanically swaged, Flemish eye splice for secure and reliable termination, and its capacities are clearly displayed on a TUFF-TAG™. It meets stringent OSHA 1910.184 and ASME B30.9 safety standards.
  • 5/8 inch diameter and 6 ft length for versatile use.
  • 6x19 IWRC EIPS construction ensures good abrasion resistance.
  • Mechanically swaged, Flemish eye splice provides secure termination.
  • Rated capacities: 7800 lb vertical, 5800 lb choker, 15600 lb basket.
  • Maximum operating temperature of 400°F.
  • Abrasion-resistant design enhances durability.
  • Meets OSHA 1910.184 and ASME B30.9 safety standards.
Catalog No. 50-005-29699 Supplier MSC Supplier No. 04813861
This eye and eye wire rope sling features a 5/8 in diameter and a 6 ft length, ideal for various lifting operations. It is constructed from 6x19 IWRC EIPS steel, providing excellent abrasion resistance and value. The sling is equipped with a mechanically swaged, Flemish eye splice for secure and reliable termination, and its capacities are clearly displayed on a TUFF-TAG™. It meets stringent OSHA 1910.184 and ASME B30.9 safety standards.
  • 5/8 inch diameter and 6 ft length for versatile use.
  • 6x19 IWRC EIPS construction ensures good abrasion resistance.
  • Mechanically swaged, Flemish eye splice provides secure termination.
  • Rated capacities: 7800 lb vertical, 5800 lb choker, 15600 lb basket.
  • Maximum operating temperature of 400°F.
  • Abrasion-resistant design enhances durability.
  • Meets OSHA 1910.184 and ASME B30.9 safety standards.