Learn More
MSC Lift-All Flat Quick Sleeve Wear Pad, Polyester, 1 ft Length, 4 in Width
Catalog No. 5000529742
Encompass
A flat quick sleeve wear pad designed for various slings, constructed from durable polyester material. It features a length of 1 foot and a width of 4 inches, suitable for protecting both chain and wire rope slings.
  • Designed for use with all slings.
  • Constructed from polyester material.
  • Overall length of 1 foot.
  • Overall width of 4 inches.
  • Compatible with chain slings with a diameter of 0.21875 in.
  • Compatible with wire ropes with a diameter of 0.5 in.
  • Features a 2 in sling width.
  • Available in gray and white.
Catalog No. 50-005-29742 Supplier MSC Supplier No. 04940375
A flat quick sleeve wear pad designed for various slings, constructed from durable polyester material. It features a length of 1 foot and a width of 4 inches, suitable for protecting both chain and wire rope slings.
  • Designed for use with all slings.
  • Constructed from polyester material.
  • Overall length of 1 foot.
  • Overall width of 4 inches.
  • Compatible with chain slings with a diameter of 0.21875 in.
  • Compatible with wire ropes with a diameter of 0.5 in.
  • Features a 2 in sling width.
  • Available in gray and white.