Learn More
MSC Lift-All Web Sling, Endless, Polyester, 1 in, 3200 lb Capacity
Catalog No. 5000534584
Encompass
This polyester endless web sling offers versatile and safe lifting for industrial applications. Featuring a 1-ply construction, 1 inch width, and a 3200 lb vertical capacity, it meets stringent safety standards.
  • Endless, Type 5 design for flexible use in various hitches.
  • Polyester construction provides enhanced load control and abrasion resistance.
  • 1-ply, 1 inch wide sling with a 6 foot length.
  • Rated for 3200 lb vertical, 2500 lb choker, and 6400 lb basket capacities.
  • Red core yarns act as a visible damage indicator.
  • Tuff-Tag ensures durable and legible product information.
  • Complies with OSHA 1910.184, ASME B30.9, and WSTDA safety standards.
  • Rotatable design minimizes wear and extends service life.
  • Allows for spreading of sling legs to improve load balance.
  • Maximum operating temperature of 200°F.
Catalog No. 50-005-34584 Supplier MSC Supplier No. 86616042
This polyester endless web sling offers versatile and safe lifting for industrial applications. Featuring a 1-ply construction, 1 inch width, and a 3200 lb vertical capacity, it meets stringent safety standards.
  • Endless, Type 5 design for flexible use in various hitches.
  • Polyester construction provides enhanced load control and abrasion resistance.
  • 1-ply, 1 inch wide sling with a 6 foot length.
  • Rated for 3200 lb vertical, 2500 lb choker, and 6400 lb basket capacities.
  • Red core yarns act as a visible damage indicator.
  • Tuff-Tag ensures durable and legible product information.
  • Complies with OSHA 1910.184, ASME B30.9, and WSTDA safety standards.
  • Rotatable design minimizes wear and extends service life.
  • Allows for spreading of sling legs to improve load balance.
  • Maximum operating temperature of 200°F.