Learn More
MSC Lift-All Web Sling, Flat Eye & Eye, 16500 lb, Polyester
Catalog No. 5000515391
Encompass
This web sling is constructed from polyester and features a flat eye and eye design. It offers a vertical capacity of 16500 lb, making it suitable for various lifting applications.
  • Flat eye & eye, type 3 design
  • 10 ft long, 6 in wide
  • 16500 lb vertical capacity
  • 13200 lb choker capacity
  • 33000 lb basket capacity
  • 2 plies
  • Yellow color
  • Maximum operating temperature of 200°F
  • Abrasion-resistant
  • Meets WSTDA, ASME B30.9, and OSHA 1910.184 standards
Catalog No. 50-005-15391 Supplier MSC Supplier No. 85818029
This web sling is constructed from polyester and features a flat eye and eye design. It offers a vertical capacity of 16500 lb, making it suitable for various lifting applications.
  • Flat eye & eye, type 3 design
  • 10 ft long, 6 in wide
  • 16500 lb vertical capacity
  • 13200 lb choker capacity
  • 33000 lb basket capacity
  • 2 plies
  • Yellow color
  • Maximum operating temperature of 200°F
  • Abrasion-resistant
  • Meets WSTDA, ASME B30.9, and OSHA 1910.184 standards