Learn More
MSC Lift-All Web Sling, Flat Eye & Eye, 7 ft Long, 2 in Wide, 2400 lb Vertical Capacity
Catalog No. 5000529977
Encompass
This Flat Eye & Eye, Type 3 Web Sling is engineered for diverse lifting applications, providing a robust vertical capacity of 2400 lb. Constructed from durable polyester, it offers abrasion resistance, making it ideal for demanding industrial environments.
  • 1900 lb Choker capacity
  • 4800 lb Basket capacity
  • Abrasion-resistant
  • Complies with WSTDA, ASME B30.9, and OSHA 1910.184 standards
  • Maximum operating temperature of 200°F
  • Yellow color for easy identification
  • Single ply construction
  • 10 inch eye length
Catalog No. 50-005-29977 Supplier MSC Supplier No. 01136753
This Flat Eye & Eye, Type 3 Web Sling is engineered for diverse lifting applications, providing a robust vertical capacity of 2400 lb. Constructed from durable polyester, it offers abrasion resistance, making it ideal for demanding industrial environments.
  • 1900 lb Choker capacity
  • 4800 lb Basket capacity
  • Abrasion-resistant
  • Complies with WSTDA, ASME B30.9, and OSHA 1910.184 standards
  • Maximum operating temperature of 200°F
  • Yellow color for easy identification
  • Single ply construction
  • 10 inch eye length