Learn More
MSC Lift-All Web Sling, Nylon, 3 in x 10 ft, 4800 lb
Catalog No. 5000535604
Encompass
This nylon web sling is designed to minimize shock loading due to its inherent stretch. It features red core warning yarns for enhanced inspection safety and a TUFF-TAG for permanent identification as required by OSHA regulations. The yellow treatment also provides increased abrasion resistance.
  • Nylon web stretches more, which helps avoid shock loading.
  • Red core warning yarns aid the inspection process.
  • Permanent identification provided by TUFF-TAG as required by OSHA.
  • Yellow treatment resists abrasion.
Catalog No. 50-005-35604 Supplier MSC Supplier No. 76691435
This nylon web sling is designed to minimize shock loading due to its inherent stretch. It features red core warning yarns for enhanced inspection safety and a TUFF-TAG for permanent identification as required by OSHA regulations. The yellow treatment also provides increased abrasion resistance.
  • Nylon web stretches more, which helps avoid shock loading.
  • Red core warning yarns aid the inspection process.
  • Permanent identification provided by TUFF-TAG as required by OSHA.
  • Yellow treatment resists abrasion.