Learn More
MSC Lift-All Web Sling, Polyester, Twisted Eye & Eye, 3200 lb, 2 in x 10 ft
Catalog No. 5000531619
Encompass
This polyester web sling is designed for heavy-duty lifting and rigging tasks, featuring a twisted eye and eye configuration. It offers a vertical hitch capacity of 3200 lb and a basket hitch capacity of 6400 lb, making it suitable for a variety of applications. With a width of 2 inches and a length of 10 feet, this single-ply sling provides reliable performance.
  • Constructed from durable polyester material.
  • Twisted eye and eye configuration for secure attachment.
  • 3200 lb vertical hitch capacity.
  • 6400 lb basket hitch capacity.
  • Dimensions of 2 in width and 10 ft length.
  • Single-ply design.
  • Yellow color for improved visibility.
  • Heavy duty web grade for demanding environments.
  • Latex yarn coating provides additional protection.
  • Suitable for temperatures up to 200°F.
Catalog No. 50-005-31619 Supplier MSC Supplier No. 75650754
This polyester web sling is designed for heavy-duty lifting and rigging tasks, featuring a twisted eye and eye configuration. It offers a vertical hitch capacity of 3200 lb and a basket hitch capacity of 6400 lb, making it suitable for a variety of applications. With a width of 2 inches and a length of 10 feet, this single-ply sling provides reliable performance.
  • Constructed from durable polyester material.
  • Twisted eye and eye configuration for secure attachment.
  • 3200 lb vertical hitch capacity.
  • 6400 lb basket hitch capacity.
  • Dimensions of 2 in width and 10 ft length.
  • Single-ply design.
  • Yellow color for improved visibility.
  • Heavy duty web grade for demanding environments.
  • Latex yarn coating provides additional protection.
  • Suitable for temperatures up to 200°F.