Learn More
MSC Lift-All Web Sling, Twisted Eye & Eye, 16 ft, 2 in, 3200 lb Vertical Capacity
Catalog No. 5000531979
Encompass
Lift-All Twisted Eye & Eye, Type 4 Web Sling is designed for lifting applications. Constructed from durable polyester, it measures 16 feet long and 2 inches wide, offering a vertical capacity of 3200 lb. This sling is suitable for various industrial lifting tasks, ensuring reliability and safety.
  • Abrasion-resistant
  • Meets WSTDA, ASME B30.9, and OSHA 1910.184 standards
Catalog No. 50-005-31979 Supplier MSC Supplier No. 75650788
Lift-All Twisted Eye & Eye, Type 4 Web Sling is designed for lifting applications. Constructed from durable polyester, it measures 16 feet long and 2 inches wide, offering a vertical capacity of 3200 lb. This sling is suitable for various industrial lifting tasks, ensuring reliability and safety.
  • Abrasion-resistant
  • Meets WSTDA, ASME B30.9, and OSHA 1910.184 standards