Learn More
MSC Lift-All Web Sling, Twisted Eye & Eye, Polyester, 16500 lb Vertical Capacity
Catalog No. 5000527608
Encompass
The Lift-All Twisted Eye & Eye, Type 4 Web Sling is designed for various lifting applications. Constructed with 2-ply polyester, it offers durability and abrasion resistance. This sling meets WSTDA, ASME B30.9, and OSHA 1910.184 standards, ensuring safety and reliability for professional use.
  • Twisted Eye & Eye, Type 4 design
  • Two-ply polyester construction
  • 3 ft length and 6 inch width
  • 16500 lb vertical capacity
  • 13200 lb choker capacity
  • 33000 lb basket capacity
  • Abrasion-resistant
  • Meets WSTDA, ASME B30.9, and OSHA 1910.184 standards
Catalog No. 50-005-27608 Supplier MSC Supplier No. 85820900
The Lift-All Twisted Eye & Eye, Type 4 Web Sling is designed for various lifting applications. Constructed with 2-ply polyester, it offers durability and abrasion resistance. This sling meets WSTDA, ASME B30.9, and OSHA 1910.184 standards, ensuring safety and reliability for professional use.
  • Twisted Eye & Eye, Type 4 design
  • Two-ply polyester construction
  • 3 ft length and 6 inch width
  • 16500 lb vertical capacity
  • 13200 lb choker capacity
  • 33000 lb basket capacity
  • Abrasion-resistant
  • Meets WSTDA, ASME B30.9, and OSHA 1910.184 standards