Learn More
MSC Lift-All Webmaster 1200 Sling, Polyester, 1-Ply, Eye & Eye
Catalog No. 5000534315
Encompass
The Webmaster® 1200 Slings are economical web slings designed for less frequent use. They feature a red core yarn warning system for safety and a Tuff-Tag™ for traceability and OSHA information. They are made of polyester.
  • Red core yarn warning system aids in the inspection process.
  • Proven reliability.
  • Tuff-Tag™ provides serial numbered identification for traceability.
  • Economical option for less frequent use.
  • Yellow treatment for abrasion resistance and extended sling life.
  • Tuff-Tag™ provides required OSHA information for the life of the sling.
Catalog No. 50-005-34315 Supplier MSC Supplier No. 01137082
The Webmaster® 1200 Slings are economical web slings designed for less frequent use. They feature a red core yarn warning system for safety and a Tuff-Tag™ for traceability and OSHA information. They are made of polyester.
  • Red core yarn warning system aids in the inspection process.
  • Proven reliability.
  • Tuff-Tag™ provides serial numbered identification for traceability.
  • Economical option for less frequent use.
  • Yellow treatment for abrasion resistance and extended sling life.
  • Tuff-Tag™ provides required OSHA information for the life of the sling.