Learn More
MSC Lift-All Wire Mesh Sling, 10 Gauge, 6 Inch Width, 10 Foot Length, Triangle & Choker
Catalog No. 5000512994
Encompass
Engineered for demanding lifting operations, this heavy-duty wire mesh sling is constructed from galvanized high-tensile steel mesh and features zinc-plated alloy steel end fittings. It offers robust resistance to corrosion, abrasion, and cutting. Each sling is proof-tested and certified, ensuring reliability and safety, with a wide-bearing area to distribute loads and minimize damage.
  • Heavy-duty 10 gauge galvanized steel mesh construction.
  • Zinc-plated alloy steel end fittings for enhanced durability.
  • Permanently stamped with capacity and serial number for traceability.
  • Proof-tested and certified to ensure safety and performance.
  • Resistant to abrasion and cutting for extended service life.
  • Designed with low stretch and a wide-bearing area to protect loads.
  • Repairable and re-certifiable to maintain original specifications.
Catalog No. 50-005-12994 Supplier MSC Supplier No. 89751424
Engineered for demanding lifting operations, this heavy-duty wire mesh sling is constructed from galvanized high-tensile steel mesh and features zinc-plated alloy steel end fittings. It offers robust resistance to corrosion, abrasion, and cutting. Each sling is proof-tested and certified, ensuring reliability and safety, with a wide-bearing area to distribute loads and minimize damage.
  • Heavy-duty 10 gauge galvanized steel mesh construction.
  • Zinc-plated alloy steel end fittings for enhanced durability.
  • Permanently stamped with capacity and serial number for traceability.
  • Proof-tested and certified to ensure safety and performance.
  • Resistant to abrasion and cutting for extended service life.
  • Designed with low stretch and a wide-bearing area to protect loads.
  • Repairable and re-certifiable to maintain original specifications.