Learn More
MSC Lift America Endless Sling, 1 in, 2 ft, 3,200 lb Vertical Capacity, Polyester
Catalog No. 5000536390
Encompass
This endless sling is engineered for versatile lifting applications. Constructed from durable polyester, it features a 1 in width and 2 ft length, offering a vertical capacity of 3,200 lb. This Type 5 sling is ideal for general-purpose lifting tasks, conforming to ASME B30.9 standards.
  • Endless sling type
  • 1 in width
  • 2 ft length
  • 3,200 lb vertical capacity
  • 2,500 lb choker capacity
  • 6,400 lb basket capacity
  • Polyester material
  • Yellow color
  • Single ply construction
  • Meets ASME B30.9 standards
Catalog No. 50-005-36390 Supplier MSC Supplier No. 16325888
This endless sling is engineered for versatile lifting applications. Constructed from durable polyester, it features a 1 in width and 2 ft length, offering a vertical capacity of 3,200 lb. This Type 5 sling is ideal for general-purpose lifting tasks, conforming to ASME B30.9 standards.
  • Endless sling type
  • 1 in width
  • 2 ft length
  • 3,200 lb vertical capacity
  • 2,500 lb choker capacity
  • 6,400 lb basket capacity
  • Polyester material
  • Yellow color
  • Single ply construction
  • Meets ASME B30.9 standards