Learn More
Grainger Lift Chain American Standard
Catalog No. 502765935
Encompass_Preferred
The Tripp Lite 16U SmartRack Wall Mount Rack Enclosure Cabinet is a 16U wall-mount rack that provides a sturdy, secure rack enclosure for storing EIA-standard 19-inch rack equipment in network closets, back offices and other space-constrained areas. Front and rear pairs of vertical rails have square mounting holes that allow easy equipment mounting at any height. Adjustable mounting rails support equipment up to 20.5 inches deep. Locking steel cabinet with vented sides provides security while allowing air circulation. Cabinet meets EIA-310-D standards and has a weight capacity of 200 lbs.
Catalog No. 50-276-5935 Supplier Grainger Supplier No. 16U030
The Tripp Lite 16U SmartRack Wall Mount Rack Enclosure Cabinet is a 16U wall-mount rack that provides a sturdy, secure rack enclosure for storing EIA-standard 19-inch rack equipment in network closets, back offices and other space-constrained areas. Front and rear pairs of vertical rails have square mounting holes that allow easy equipment mounting at any height. Adjustable mounting rails support equipment up to 20.5 inches deep. Locking steel cabinet with vented sides provides security while allowing air circulation. Cabinet meets EIA-310-D standards and has a weight capacity of 200 lbs.