Learn More
Elma™ Lift-off Lids for Elmasonic Xtra ST Ultrasonic Cleaner
Description
- Stainless steel construction
- Helps to reduce noise and evaporation, serves as drip trays
- Insulated for sound reduction
Specifications
Specifications
| Product Type | Lid |
| For Use With (Equipment) | Ultrasonic Cleaner, Elmasonic Xtra ST 2500 H |
| Style | Lift-Off Lid |
| For Use With (Application) | Helps to Reduce Noise and Evaporation, Serves as Drip Trays |
| Material | Stainless Steel |