Learn More
Uline Battery Operated Lift Table - Standard, 1,650 lb, 40 x 20"
Catalog No. NC2865738
Encompass
Battery powered lift table can go anywhere at any time.Raise loads up to 5x faster with the push of a button.Hefty 1HP DC motor easily lifts heavy items.100 lifts per charge: 24-volt, 30 Ah maintenance-free battery with built-in charger.Polyurethane casters: 2 swivel with brakes, 2 rigid.
Catalog No. NC2865738 Supplier Uline Supplier No. H3768
Battery powered lift table can go anywhere at any time.Raise loads up to 5x faster with the push of a button.Hefty 1HP DC motor easily lifts heavy items.100 lifts per charge: 24-volt, 30 Ah maintenance-free battery with built-in charger.Polyurethane casters: 2 swivel with brakes, 2 rigid.