Learn More
Julabo USA Inc Lift-Up Bath cover for PURA 10 water bath, transparent
SDP
Catalog No. 501904631
Encompass_Preferred
Lift-up, transparent bath covers for JULABO PURA water baths. Necessary for operation in the PURA bath tank to temperatures >90 °C. Bath cover made from Plexiglas. Easy installation with a screwdriver.
Catalog No. 50-190-4631 Supplier Julabo USA Inc Supplier No. 9970581
Lift-up, transparent bath covers for JULABO PURA water baths. Necessary for operation in the PURA bath tank to temperatures >90 °C. Bath cover made from Plexiglas. Easy installation with a screwdriver.