Learn More
Julabo USA Inc Each, Lift-up plastic bath lid for SW22/SW23 shaking water baths
SDP
Catalog No. 507534106
Encompass_Preferred
Bath Accessories, Julabo, Lift-up Makrolon cover WxLxH: (51x35x16 cm): Prevent liquid losses due to evaporation, protects samples from contamination to +80°C, for SW22 and SW23 shaking water baths.
This item is not returnable. View return policy
Catalog No. 50-753-4106 Supplier Julabo USA Inc Supplier No. 8970288
Bath Accessories, Julabo, Lift-up Makrolon cover WxLxH: (51x35x16 cm): Prevent liquid losses due to evaporation, protects samples from contamination to +80°C, for SW22 and SW23 shaking water baths.