Learn More
Research Products International Corp Lift-Away Concentrated Radioactivity Decontaminant, 100 Milliliters, 6 per Pack
SDP
Catalog No. 50212990
Encompass_Preferred
Lift-Away provides the most effective and convenient solution for dealing with radioactive contamination. Non-hazardous, biodegradable formula is safe to use on all surfaces including skin, glass, plastic, metal, fabrics, walls, paint, floors, tile, rubber, wood, instruments and utensils. Ideal general purpose cleaner for laboratory glassware, Also removes stubborn contaminants such as soil, grease, dried blood, resins and tissue.
Easy to use, simply spray directly on area to be cleaned or apply with a cloth, sponge or brush. Lift-Away can be diluted with up to 10 parts water.
This item is not returnable. View return policy
Catalog No. 50-212-990 Supplier Research Products International Corp Supplier No. 114005
Lift-Away provides the most effective and convenient solution for dealing with radioactive contamination. Non-hazardous, biodegradable formula is safe to use on all surfaces including skin, glass, plastic, metal, fabrics, walls, paint, floors, tile, rubber, wood, instruments and utensils. Ideal general purpose cleaner for laboratory glassware, Also removes stubborn contaminants such as soil, grease, dried blood, resins and tissue.
Easy to use, simply spray directly on area to be cleaned or apply with a cloth, sponge or brush. Lift-Away can be diluted with up to 10 parts water.