Learn More
MSC Liftex Web Sling, Endless, 8 ft, 6400 lb Basket Capacity, Nylon
Catalog No. 5000528490
Encompass
The Liftex Endless, Type 5 Web Sling is constructed from durable nylon, designed for high-capacity lifting operations. This 8 ft long and 1 in wide sling offers a 6400 lb basket capacity, ensuring reliable and safe material handling. It is engineered to meet or exceed industry standards for strength and safety, providing a premium edge for extended sling life and increased safety.
  • Endless, type 5 design for versatile lifting configurations.
  • Manufactured from durable nylon material.
  • 8 ft length and 1 inch width for adaptable use.
  • Features a 6400 lb basket capacity.
  • Equipped with premium edge and tag for extended service life and enhanced safety.
  • Adheres to or surpasses OSHA, ASME B30.9, and WSTDA safety and strength standards.
  • Single ply construction.
  • Yellow color for easy identification.
Catalog No. 50-005-28490 Supplier MSC Supplier No. 08924441
The Liftex Endless, Type 5 Web Sling is constructed from durable nylon, designed for high-capacity lifting operations. This 8 ft long and 1 in wide sling offers a 6400 lb basket capacity, ensuring reliable and safe material handling. It is engineered to meet or exceed industry standards for strength and safety, providing a premium edge for extended sling life and increased safety.
  • Endless, type 5 design for versatile lifting configurations.
  • Manufactured from durable nylon material.
  • 8 ft length and 1 inch width for adaptable use.
  • Features a 6400 lb basket capacity.
  • Equipped with premium edge and tag for extended service life and enhanced safety.
  • Adheres to or surpasses OSHA, ASME B30.9, and WSTDA safety and strength standards.
  • Single ply construction.
  • Yellow color for easy identification.