Learn More
MSC Liftex Web Sling, Polyester, 1 inch, 6 feet, 3200 lb Basket Capacity
Supplier: MSC 59619809
This durable web sling is engineered for heavy-duty lifting applications, featuring a flat eye & eye design for versatile use. Constructed with Pro-Edge technology, it offers enhanced resistance to wear and abrasion, making it 50% tougher than standard webbing. Its high-tenacity polyester fibers ensure robust support and long-lasting performance.
- 50% tougher than standard webbing for enhanced durability
- High-tenacity polyester fibers provide long-lasting wear and abrasion resistance
- Flat eye & eye configuration supports various lifting methods
- Vertical capacity of 1600 lb, choker capacity of 1250 lb, and basket capacity of 3200 lb
- Manufactured in the U.S.A.
- Meets ASME B30.9 and WSTDA safety standards
- Equipped with a durable tag for maximized information life