Learn More
Grainger LiftOff Hinge Assembly 2 25/64 x2 5/16in
Catalog No. 502763964
Encompass_Preferred
The 80/20 Hinge 2-25/64 in x 2-5/16 in is a heavy-duty hinge made from aluminum with a black anodized finish. It has a 2-25/64 inch height, 2-5/16 inch width, and 0.375 inch diameter. The hinge features a self-lubricating acetal bearing, stainless steel pin, and can support up to 300 lbs per pair. It is designed for use in industrial machinery, automation equipment, and anywhere heavy duty hinges are required. The hinge has a temperature range of -20°F to 250°F.
Catalog No. 50-276-3964 Supplier Grainger Supplier No. 16U360
The 80/20 Hinge 2-25/64 in x 2-5/16 in is a heavy-duty hinge made from aluminum with a black anodized finish. It has a 2-25/64 inch height, 2-5/16 inch width, and 0.375 inch diameter. The hinge features a self-lubricating acetal bearing, stainless steel pin, and can support up to 300 lbs per pair. It is designed for use in industrial machinery, automation equipment, and anywhere heavy duty hinges are required. The hinge has a temperature range of -20°F to 250°F.