Learn More
Aviva Systems Biology Corp Lig3 Antibody - middle region (ARP54959_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54959_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Lig3. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: Lig3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-983