Learn More
Aviva Systems Biology Corp LIG4 Antibody - N-terminal region (ARP34122_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34122_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against LIG4. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human LIG4. Alternate names: ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55257