Learn More
Med Vet International Liga Fiba Interference Screw 3.5mm x 18mm, J0444R
Catalog No. 5000581975
Encompass_Preferred
An alternative to using toggle buttons with LigaFiba is using these interference screws. They give a low profile alternative to tying over a button. The screw has tapered soft profile thread which jams the LigaFiba into the bone tunnel without damaging it. The screw may be partially introduced then the joint checked for stability. If satisfied, the screw is driven to lock the suture. Hex Head. 3.5mm stainless. Uses J0412b hex head screw driver for 3.5mm screws.
Catalog No. 50-005-81975 Supplier Med Vet International Supplier No. JOR444R
An alternative to using toggle buttons with LigaFiba is using these interference screws. They give a low profile alternative to tying over a button. The screw has tapered soft profile thread which jams the LigaFiba into the bone tunnel without damaging it. The screw may be partially introduced then the joint checked for stability. If satisfied, the screw is driven to lock the suture. Hex Head. 3.5mm stainless. Uses J0412b hex head screw driver for 3.5mm screws.