Learn More
Med Vet International LigaFiba Braided Polyethylene Fiber (Material Only), J0444A, 150lbs, 500mm, Each
Catalog No. 5000581902
Encompass_Preferred
Monofilament nylon has been the standard suture material for the lateral suture technique for repair of CCL damage. Newer materials are made of braided polyethylene fiber and offer several advantages over nylon.Significantly stronger (up to 15 times)Better flex fatigue resistance and abrasionKnots tie easier, this makes LigaFiba suture material an ideal material for ligament and tendon repair. Its available packages are:LigaFiba suture material onlyProvided with attached cruciate needle and straight needleLigaFiba with toggle buttons in bone tunnel procedures in both cruciate and hip repairs
Catalog No. 50-005-81902 Supplier Med Vet International Supplier No. JOR444A
Monofilament nylon has been the standard suture material for the lateral suture technique for repair of CCL damage. Newer materials are made of braided polyethylene fiber and offer several advantages over nylon.Significantly stronger (up to 15 times)Better flex fatigue resistance and abrasionKnots tie easier, this makes LigaFiba suture material an ideal material for ligament and tendon repair. Its available packages are:LigaFiba suture material onlyProvided with attached cruciate needle and straight needleLigaFiba with toggle buttons in bone tunnel procedures in both cruciate and hip repairs