Learn More
Molecular Dimensions The Ligand-Friendly Screen
SDP
Catalog No. NC1604552
Encompass
The Ligand-Friendly Screen (LFS)1 has been one of the two primary coarse screens of the Structural Genomics Consortium (SGC) in Oxford since 2004; at last count, it had produced 190 novel structures over 10 years.
Catalog No. NC1604552 Supplier Molecular Dimensions Supplier No. MD1122
The Ligand-Friendly Screen (LFS)1 has been one of the two primary coarse screens of the Structural Genomics Consortium (SGC) in Oxford since 2004; at last count, it had produced 190 novel structures over 10 years.