Learn More
Fine Science Tools Ligation Aids, Hole Diameter: 0.4 mm, 10.5 cm
SDP
Catalog No. NC9749346
Encompass
Product: Ligation Aid - 10.5cm/0.4mm Hole Diameter/ Hole Diameter: 0.4mm/ Alloy: Stainless Steel10.5cm/ Feature: Ultrafine Each
Catalog No. NC9749346 Supplier Fine Science Tools Supplier No. 1806212
Product: Ligation Aid - 10.5cm/0.4mm Hole Diameter/ Hole Diameter: 0.4mm/ Alloy: Stainless Steel10.5cm/ Feature: Ultrafine Each