Learn More
Cayman Chemical LIGHCD258 EXrAClular Dom 100ug
SDP
Catalog No. 502947266
Encompass_Preferred
Source: Active recombinant N-terminal human IgG1 Fc-tagged LIGHT expressed in HEK293 cells • Amino acids: 74-240 • MW: 44.9 kDa
Catalog No. 50-294-7266 Supplier Cayman Chemical Supplier No. 31841100 UG
Source: Active recombinant N-terminal human IgG1 Fc-tagged LIGHT expressed in HEK293 cells • Amino acids: 74-240 • MW: 44.9 kDa