Learn More
CELLTREAT Scientific Products Light Absorbing Sealing Film, Black, Non-sterile
SDP
Catalog No. 5000144324
Encompass_Preferred
Black and white film is suitable for application on the top or bottom of black and white multiple well platesEasily cover clear bottom black or white plates to prepare for top-reading analytical equipmentBlack film absorbs stray light and fluorescence, dampening background noise and improving signal clarityWhite film reflects weak light and fluorescent signals to enhance output readingsPlace black film on top of black plates to fully shield light-sensitive samples
Catalog No. 50-001-44324 Supplier CELLTREAT Scientific Products Supplier No. 229141
Black and white film is suitable for application on the top or bottom of black and white multiple well platesEasily cover clear bottom black or white plates to prepare for top-reading analytical equipmentBlack film absorbs stray light and fluorescence, dampening background noise and improving signal clarityWhite film reflects weak light and fluorescent signals to enhance output readingsPlace black film on top of black plates to fully shield light-sensitive samples